Learn More
Trajan Scientific and Medical™ Needle cleaning kit includes stylet wires and tweezers SGE
Description
- Effective cleaning of syringe needle
- Kit available as well as replacement wires
Specifications
Specifications
| Includes | Tube of Stylet Wires 150mm x 0.10mm O.D., Tube of Stylet Wires 150mm x 0.15mm O.D., Tube of Stylet Wires 150mm x 0.20mm O.D., 20mL Cleaning Solution Concentrate, Tweezer |
| Product Type | Needle Cleaning Kit |