Learn More
VELP Scientifica NDA 702 Dumas Nitrogen Analyzer
Shop All Velp Products
Click to view available options
Type:
Elemental Analyzer
Unit Size:
Each
Description
FAST ANALYSIS
- The fully automatic NDA 702 elemental analyzer perform analysis in 3-4 minutes. The quick start and sleeping mode ensure fast operation set up with possibility of leak test per zone.
THE HIGHEST PRECISION
- The LoGas™ TCD (Thermal Conductivity Detector) designed by VELP guarantees unmatched precision with the unrivaled LOD of 0,001 mgN and low standard deviation.
MAXIMUM SAFETY
- The NDA 702 is equipped with state-of-the-art safety features that interrupt the analysis in case of irregularities. Immediate notification and remote stop via VELP Ermes Cloud Platform ensures safe unattended operations.
EFFICIENCY
- The fully automatic and reliable combustion method of the NDA 702 together with DriStep™ water trap guarantee moderate cost. The 30-positions disc electronic autosampler expandable with extra 3 discs ensures maximum productivity and uptime.
VERSATILITY
- The NDA 702 works with solid, liquid and doughy samples of up to 1 gram. Seamless carrier gas selection between Helium and Argon without hardware changes and whit no memory effect.
SMART INSTRUMENT
- The NDA 702 N/protein analyzer is easy to use and does not require skilled operator. The user-friendly DUMASoft™ software and VELP Ermes connection will deliver a new user experience and smart workflows.
Specifications
Specifications
| Type | Elemental Analyzer |
| Detector Type | Thermal Conductivity Detector |
| For Use With (Application) | Food Safety, Environmental Science, Agronomy |