missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NBMPR

Product Code. 18055472
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18055472 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18055472 Supplier Tocris Bioscience™ Supplier No. 2924/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Equilibrative nucleoside transporter 1 (ENT1) inhibitor

Equilibrative nucleoside transporter 1 (ENT1) inhibitor (Ki values are 0.4 and 2800 nM for hENT1 and hENT2 respectively).
TRUSTED_SUSTAINABILITY

Specifications

CAS 38048-32-7
Quantity 50 mg
Molecular Formula C17H17N5O6S
Formula Weight 419.41
Purity >99%
Molecular Weight (g/mol) 419.412
Chemical Name or Material NBMPR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.