Learn More
Saint Gobain Life Sciences Natural Rubber Stoppers
Product Code. 11734538
Click to view available options
Diameter (Metric) Bottom:
4 mm
5 mm
8 mm
10.5 mm
15 mm
17 mm
18 mm
21 mm
29 mm
34 mm
36 mm
41 mm
47 mm
Quantity:
10/Pk.
100/ck.
20/Pk.
5/Pk.
50/Pk.
Unit Size:
Pack of 10
Pack of 100
Pack of 20
Pack of 5
Pack of 50
Product Code. 11734538
Brand: Saint Gobain Life Sciences 20304910
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Made from elastic natural rubber, these stoppers are flexible and provide a complete seal and good adhesion. Saint Gobain Life Sciences Performance Plastics™ Natural Rubber Stoppers withstand chemical agents in normal laboratory use. Ideal for use with pipets, vials, bottles, flasks or balloons.
  • Temperature resistance from -35 to +70°C (120°C in humid conditions)
  • DIN Standard 18271
  • Gray-blue color
TRUSTED_SUSTAINABILITY
Specifications
Height (Metric) 40mm
Quantity 10/Pk.
Color Blue
Diameter (Metric) Bottom 41 mm
Diameter (Metric) Top 49 mm
Material Natural Rubber
Product Type Stopper
Stopper Number 49D