Learn More
BRAND™ Natural Rubber Stoppers
Product Code. 15281439 Shop All Brand Products
Change view
Click to view available options
Diameter (Metric) Top:
12 mm
14 mm
22 mm
55 mm
Unit Size:
Each
Pack of 25
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Top unitSize
15281439 14 mm Pack of 25
15211449 22 mm Pack of 25
10420293 55 mm Each
15271439 12 mm Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15281439 Supplier BRAND™ Supplier No. 144384
Only null left
Add to basket
Add to basket
Protects liquids from outside contaminants. Provides a secure seal.
  • Natural rubber construction
  • Red color
  • Supplied in packs of 25
TRUSTED_SUSTAINABILITY
Specifications
Height (Metric) 20 mm
Quantity 25/PK
Color Red
Diameter (Metric) Bottom 11 mm
Diameter (Metric) Top 14 mm
Material Natural Rubber