Learn More
Ansell Edmont™ Marigold™ Natural Rubber Industrial Protective Glove
Product Code. 12931115
Change view
Click to view available options
Size:
7.5
8.5
10.5
Unit Size:
12 pairs
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12931115 8.5 12 pairs
15170259 10.5 Each
15120154 7.5 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12931115 Supplier Ansell Edmont™ Supplier No. A005085
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Ideal for use with heavy duty protection.
  • Chemical
  • Unlined black natural rubber latex glove
  • Chlorinated to harden and cleanse the surface of the glove
  • Beaded cuff for tear resistance and easy donning
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Exterior Finish Smooth
Material Rubber Latex
Size 8.5
Glove Cuff Style Beaded
Resistant To Chemicals