Learn More
Saint-Gobain Natural Rubber Agitator
Product Code. 11704638
Click to view available options
Diameter (Metric):
6 mm
Unit Size:
Pack of 100
Product Code. 11704638
Brand: Saint-Gobain 101029
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Saint Gobain Performance Plastics™ Natural Rubber Agitator is a policemen style stirrers fit easily on any glass or plastic cane.
  • Do not break
  • Do not fissure glassware
TRUSTED_SUSTAINABILITY
Specifications
Description Natural Rubber
Diameter (Metric) 6 mm
Quantity 100/Pk.