Learn More
Memmert™ Natural Convection Hot Air Sterilizer with SingleDISPLAY
Product Code. 12805853 Shop All Memmert Products
Change view
Click to view available options
Capacity (Metric):
32 L
53 L
74 L
108 L
161 L
256 L
449 L
749 L
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12805853 161 L Each
12895843 108 L Each
12815853 256 L Each
12875843 32 L Each
12825853 449 L Each
12865843 53 L Each
12885843 74 L Each
12835853 749 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 12805853 Supplier Memmert™ Supplier No. SN160
Support a Safer, More Efficient Laboratory
Use promo code 28026
View all promotions
 Restricted Item
Estimated Shipment: 11-11-2026
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Sterilize a variety of containers and instruments with this hot air sterilizer. Memmert™ Natural Convection Hot Air Sterilizers with SingleDISPLAY allow adjustment of pre-heated fresh air admixture by air flap control in 10% steps. Safer than disinfection.
  • German, English, Spanish, French, Polish, Czech, Hungarian language settings
  • SingleDISPLAY provides adaptive multifunctional digital PID-microprocessor controller with high-definition TFT-color display
  • Digital backwards counter with target time setting, adjustable from 1 minute to 99 days
  • Process time does not start until the set temperature is reached
  • Three freely selectable temperature values
  • Adjust temperature (Celsius or Fahrenheit), air flap position, program time, time zones, summertime/wintertime
Specifications
Certifications/Compliance CE, cUL, DVE, EAC
Capacity (Metric) 161 L
Type Natural Convection Hot Air Sterilizer
Display TFT
For Use With (Application) Applied for sterilisation of medical materials, complete killing of highly resistant microorganisms, special application of depyrogenisation
Includes Works Calibration Certificate
Material Stainless Steel
Model SN160
Dimensions (L x W x H) 745 x 1104 x 584 mm
Length (Metric) 584 mm
Width (Metric) 745 mm
Width (Metric) Interior 560 mm
Height (Metric) 1104 mm
Height (Metric) Interior 720 mm
Depth (Metric) 584 mm
Depth (Metric) Interior 400 mm
Shipping Weight (Metric) 96 kg
Electrical Requirements 115/230 V 50/60 Hz
Voltage 115/230 V
Wattage 1800/3200 W
Frequency 50/60 Hz