Learn More
JT Baker Sodium Hydrogen Phosphate Anhydrous BAKER, 500g
Brand:  JT Baker 0306.0500
Sodium Hydrogen Phosphate Anhydrous BAKER, 500g
Product Code. 10010400
  • ONLINE EXCLUSIVE PRICE
    98.00€
    80.74€ / 500g
    Save 17.26€
    17% Off
 Restricted Item
Estimated Shipment: 06-11-2026