Learn More
Thermo Scientific™ National Unassembled Certified™ Vial Kits
Click to view available options
Volume (Metric):
1 mL
4 mL
Closure Material:
Bonded PTFE, Silicone
Polyethylene
Closure Type:
Screw Cap
SepCap
Color:
Amber
Unit Size:
Pack of 100
Pack of 200
Description
Thermo Scientific™ National Unassembled Certified Vial Kits provide extra assurance that time after time, the product upholds rigorous manufacturing standards.
- Fully lot tested for HPLC and GC analysis
- Certificate of Conformance in every pack
- Bonded caps to prevent septa push-through
- Clear Type 1, 33 expansion borosilicate glass or amber Type 2, 51 expansion borosilicate glass
- Available with or without ID patch
- Fixed micro vials or inserts for limited-volume sampling
Specifications
Specifications
| Description | Certified 4mL Amber Glass Screw-top Vial Kit, 13-425 Screw Thread |
| Color | Amber |
| Material | PTFE, Silicone |
| Volume (Metric) | 4 mL |
| Septum | PTFE/Silicone |
| Closure Material | Bonded PTFE, Silicone |
| Closure Type | Screw Cap |
| Closure Size | 13-425 |