Learn More
Thermo Scientific™ National Target DP™ Septa
Product Code. 12428716
Click to view available options
Color:
Ivory, Red
Unit Size:
Pack of 100
Product Code. 12428716
Brand: Thermo Scientific™ C400057
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Thermo Scientific™ National Target DP Septa are precision cut to form a uniform, evaporation-resistant seal.
Thermo Scientific™ National Target DP Septa are precision cut to form a uniform, evaporation-resistant seal.
TRUSTED_SUSTAINABILITY
Specifications
Color Ivory, Red
Material PTFE, Rubber
Septum Ivory
Closure Size 9 mm