Learn More
Thermo Scientific™ National Silanized Sample Vials
Product Code. 12478686
Click to view available options
Volume (Metric):
8 mL
12 mL
40 mL
Unit Size:
Pack of 100
Pack of 200
Product Code. 12478686
Brand: Thermo Scientific™ B7999S3
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Thermo Scientific™ National Silanized Sample Vials are constructed of superior quality borosilicate clear or amber glass.
Thermo Scientific™ National Silanized Sample Vials are constructed of superior quality borosilicate clear or amber glass.
TRUSTED_SUSTAINABILITY
Specifications
Description Clear; 8mL
Diameter (Metric) Outer 17 mm
Material Glass
Volume (Metric) 8 mL
Height (Metric) 60 mm
Closure Size 15-425