Learn More
Thermo Scientific™ National Screw Thread Caps with Bonded Septa
Product Code. 12413678
Click to view available options
Pre-slit:
No
Yes
Closure Size:
10-425
13-425
8-425
Unit Size:
Pack of 100
Product Code. 12413678
Brand: Thermo Scientific™ C401068A
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Effectively eliminate septa push-through with Thermo Scientific™ National Screw Thread Caps with Bonded Septa.
Effectively eliminate septa push-through with Thermo Scientific™ National Screw Thread Caps with Bonded Septa.
  • Septa and polypropylene caps bonded together for an integral system
  • Eliminates septa push-through
  • Available with choice of GPI thread sizes and preslit septa for easier thin-gauge needle penetration
TRUSTED_SUSTAINABILITY
Specifications
Description Black; Bonded Red 0.005 in. PTFE/White Silicone Septa; GPI Thread Finish: 10-425
Color White, Black
Material PTFE, Silicone, Polypropylene
Pre-slit No
Septum Bonded Red PTFE/White Silicone, 0.005 in.
Product Line National
Closure Size 10-425