Learn More
Thermo Scientific™ National Scientific PTFE/Silicone Seals
Product Code. 12420278
Click to view available options
Unit Size:
Pack of 1000
Product Code. 12420278
Brand: Thermo Scientific™ C402032AB
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
20mm dia. open seal sillicone septa supplied in packs of 1,000
  • PTFE/Silicone construction
  • 20mm dia.
  • Open seal
  • Silicone septa
  • Supplied in packs of 1,000
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Outer 20 mm
Material Silicone
Quantity 1000/Cs.