Learn More
Thermo Scientific™ Manual Crimpers and Decrimpers for Cleanrooms
Product Code. 15200379
Change view
Click to view available options
Size:
11 mm
13 mm
20 mm
Type:
Manual Crimper
Manual Decrimper
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Type unitSize
15200379 11 mm Manual Crimper Each
15277298 13 mm Manual Crimper Each
15267298 11 mm Manual Decrimper Each
15210379 13 mm Manual Decrimper Each
15220379 20 mm Manual Decrimper Each
15217308 20 mm - Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15200379 Supplier Thermo Scientific™ Supplier No. C4012100SS
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
Achieve secure, repeatable crimps or remove seals with minimized risk of damage.
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use stainless-steel Thermo Scientific™ National Manual Crimpers and Decrimpers.
Crimpers provide a reproducible, secure vial closure; decrimpers allow easy removal of aluminum seals without breakage.
  • Stainless steel construction
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
No. per Pack 1 Ea.
Type Manual Crimper
For Use With (Equipment) Aluminum Seal
Size 11 mm
For Use With (Application) Industrial Chromatography
Product Line National