Learn More
Thermo Scientific™ National Scientific Inserts for Target LoVial™ wide-opening vials
Description
With a nominal O.D. of 6mm, these Thermo Scientific™ National Inserts fit Target LoVial Wide-Opening Vials.
- Pulled point inserts are an economical choice for noncritical applications
- Nominal O.D. of 6mm to fit Target wide-opening vials
For use with:
- Target LoVial Wide-Opening vials
- Snap-It™ Wide-Opening Snap Cap vials
- Target DP™ vials
- Target Wide-Opening vials
Specifications
Specifications
| Description | 150uL Glass Conical with Top Spring |
| Volume (Metric) | 150 μL |
| Style | Insert is suspended from the rim of the vial |
| Product Line | National |
| For Use With (Equipment) | Target LoVial Wide-Opening Vials |