Learn More
Thermo Scientific™ National Crimp-Top Autosampler Vials
Product Code. 11563810 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
2.5mL; 11mm; Clear
8mm x 40mm crimp top vials, clear glass flat bottom, 1.2mL
Volume (Metric):
1.2 mL
2.5 mL
Unit Size:
Pack of 100
Pack of 125
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Volume (Metric) unitSize
11563810 8mm x 40mm crimp top vials, clear glass flat bottom, 1.2mL 1.2 mL Pack of 125
10702357 2.5mL; 11mm; Clear 2.5 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11563810 Supplier Thermo Scientific™ Supplier No. 60180501
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Thermo Scientific™ National Crimp-Top Autosampler Vials are offered as clear or amber, in a range of sizes.
Thermo Scientific™ National Crimp-Top Autosampler Vials are offered as clear or amber, in a range of sizes.
Ordering Information:
Order caps separately.
TRUSTED_SUSTAINABILITY
Specifications
Description 8mm x 40mm crimp top vials, clear glass flat bottom, 1.2mL
Volume (Metric) 1.2 mL
Volume (Metric) Working 1.2 mL
Closure Size 8 mm