Learn More
Thermo Scientific™ Assembled Target™ 10-425 Caps and Septa
Product Code. 12483658
Click to view available options
Color:
Natural
Yellow
Septum:
Integral Molded Polypropylene
Red PTFE/White Silicone
Unit Size:
Pack of 100
Pack of 1000
Product Code. 12483658
Brand: Thermo Scientific™ C401060AY
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Minimize contamination from handling with convenient Thermo Scientific™ National Assembled Target 10-425 Caps and Septa.
Minimize contamination from handling with convenient Thermo Scientific™ National Assembled Target 10-425 Caps and Septa.
  • Pre-assembled caps and septa are convenient and minimize contamination from handling
  • Storage cap has a solid top and pre-inserted PTFE disk liner
TRUSTED_SUSTAINABILITY
Specifications
Description Yellow; Septa: Red PTFE/White Silicone
Color Yellow
Material PTFE, Silicone, Polypropylene
Septum Red PTFE/White Silicone
Product Line National Assembled Target
Type Screw Cap
Closure Size 10 mm
For Use With (Equipment) 10-425 Screw Thread Vials