Learn More
Whirl-Pak™ Stand-Up Sample Bags
Product Code. 11716873 Shop All Whirl-Pak Products
Change view
Click to view available options
Dimensions (L x W):
18.5 x 7.5 cm (7.25 x 3 in.)
23 x 11.5 cm (9 x 4.5 in.)
23 x 15 cm (9 x 6 in.)
38 x 15 cm (15 x 6 in.)
38 x 19 cm (15 x 7.5 in.)
Thickness (English):
2.50 mil
3 mil
4 mil
Capacity (Metric):
118 mL
532 mL
710 mL
1.24 L
2 L
Unit Size:
Pack of 250
Pack of 500
Pack of 6000
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Thickness (English) Capacity (Metric) Unit Size
11716873 18.5 x 7.5 cm (7.25 x 3 in.) 2.50 mil 118 mL Pack of 500
30281139 18.5 x 7.5 cm (7.25 x 3 in.) 2.50 mil 118 mL Pack of 6000
11726873 23 x 11.5 cm (9 x 4.5 in.) 3 mil 532 mL Pack of 500
11766965 23 x 15 cm (9 x 6 in.) 3 mil 710 mL Pack of 500
11781796 38 x 15 cm (15 x 6 in.) 4 mil 1.24 L Pack of 250
11791796 38 x 19 cm (15 x 7.5 in.) 4 mil 2 L Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11716873 Supplier Whirl-Pak™ Supplier No. B01364WA
  • ONLINE EXCLUSIVE PRICE
    264.00€
    85.55€ / Pack of 500
    Save 178.45€
    67% Off
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Stand-up bags make it very easy to add material to the bag and to remove sample for testing or analysis. The flat bottom ensures stability and the wire tabs hold the top open for easy access.
  • Paper-like finish virtually eliminates ink smearing
  • Surface that will accept a variety of writing instruments
  • Sterilized after manufacturing
Specifications
Description Whirl-Pak™ Stand-Up Bag
Autoclavable Not Autoclavable
Closure Type Tape and Wire
Color Clear with White Block
Disposable Single-use
Material Polyethylene
Sterility Sterile
Fill Volume 4 oz.
Latex Free Yes
No. per Pack 500/Pk.
Outer Packaging Cardboard
Protective Inner Packaging Plastic
Quantity 500/Pk.
Sterilization Method Ethylene Oxide Gas
Temperature Range (English) -320°F to +180°F
Temperature Range (Metric) -196°C to +82°C
Thickness (English) 2.50 mil
Dimensions (L x W) 18.5 x 7.5 cm (7.25 x 3 in.)
Length (English) 7.25 in.
Width (English) 3 in.
Product Type Stand-up Bag
Capacity (English) 4 oz.
Capacity (Metric) 118 mL
Length (Metric) 18 cm
Width (Metric) 7.5 cm
Thickness (Metric) 0.064 mm