Learn More
Naphtho[2,3-c]furan-1,3-dione, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 716-39-2 |
| Molecular Formula | C12H6O3 |
| MDL Number | MFCD00059091 |
| Quantity | 100 g |
| SMILES | O=C1OC(=O)C2=CC3=C(C=CC=C3)C=C12 |
| Molecular Weight (g/mol) | 198.17 |
| Percent Purity | 95% |
| Chemical Name or Material | Naphtho[2,3-c]furan-1,3-dione |