Learn More
Implen™ NanoPhotometer™ IQ/OQ Documentation and Standard Solution
Product Code. 16658323 Shop All Implen Products
Change view
Click to view available options
For Use With (Equipment):
NanoPhotometer N120
NanoPhotometer N60
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
16658323 NanoPhotometer N120 Each
15533214 NanoPhotometer N60 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16658323 Supplier Implen™ Supplier No. N120Q
 Restricted Item
Estimated Shipment: 22-10-2026
In compliance with international standards in regulated environments, documented verification is provided with Installation Qualification and Operation Qualification (IQ/OQ) Package Conforming Data to verify that instruments are working according to specifications in case of an audit.
  • Documentation provided on a USB flash drive as an Excel file
  • Includes NanoPhotometer™ appropriate standard solution (N-568-S or N-120-S)
Specifications
For Use With (Equipment) NanoPhotometer N120
Certifications/Compliance According to Internal SOP
Calibration Type Self-calibration
Product Type IQ/OQ Package Conforming Data
Product Line NanoPhotometer
Includes IQ/OQ Documentation and Standard Solution (N-120-S)