Learn More
Thermo Scientific™ NanoDrop Ultra Acclaro Pro Bundles
Shop All Thermo Scientific ProductsDescription
The Thermo Scientific™ NanoDrop™ Ultra Spectrophotometers and Fluorometers represent the next generation of microvolume detection. This cutting-edge instrument delivers exceptional accuracy and sensitivity for nucleic acid and protein analysis with sample volumes as low as 1.0 μL. The NanoDrop Ultra is designed to operate seamlessly within workflows across a wide range of fields, including genomics, proteomics, molecular biology, biotechnology, pharmaceutical, and life sciences research. It provides unparalleled precision, efficiency, and reliability in one compact device. These bundles include the purchase of one of the NanoDrop Ultra instruments with an additional factory calibration unlocking the Acclaro Pro software.
Avoid costly setbacks in your workflow with an understanding of sample quality like never before using the Thermo Scientific™ NanoDrop™ Ultra Microvolume UV-Vis Spectrophotometers Fluorometers. Innovative Thermo Scientific™ Acclaro™ Sample Intelligence technology is built into every instrument for improved measurement accuracy and contaminant identification. Quantify and qualify DNA, RNA, and protein samples in seconds with only 1–2 μL, and obtain full spectral data before you decide to use samples in downstream applications. Cuvette compatible models are available for additional flexibility.
Obtain the most complete information about the concentration and quality of your DNA or RNA samples by making absorbance or fluorescence measurements on the same instrument. The NanoDrop Ultra FL and NanoDrop UltraC FL are capable of both microvolume absorbance and fluorescence measurements, thanks to the inclusion of red and blue, solid state light emitting diodes (LEDs).
- High sensitivity — More sensitive than absorbance — Based quantification with the ability to detect down to 0.05 ng/μL dsDNA or RNA user account access.
- Sample prep calculator — Integrated reagent calculator determines amount of dye and buffer needed.
Streamlined workflow
- Fully integrated qPCR recipe calculator — Use the built–in library of kits, or add your favorite, to let the software run the math for you. The qPCR recipe calculator suggests how to prepare the reaction based on your sample's concentration.
- Auto Measure and Auto Blank functions — Multi-sample processing can be streamlined with instant measurements that occur when the pedestal arm is down. These functions can be turned ON or OFF with a single tap on the screen.
Advanced connectivity for data storage, printing and sharing
- Export to the cloud — Export data directly to popular cloud storage services like Microsoft OneDrive, Google Drive, and Thermo Fisher Connect.
- Direct LIMS integration — Send data directly into your LIMS with the assistance of an integrated RESTful API.
- Easy data–to–printer options — Print your results directly from the NanoDrop Ultra instrument via Wi-Fi, Ethernet or USB printer connection; print exported data from your PC using the Nanodrop Ultra PC Control software.
- Data Your Way — Customize and preview the information included in the data file prior to export to ensure it meets any downstream needs.
Accuracy beyond expectations with the Acclaro Pro Software
This bundle includes new versions of the most popular measurement applications, which are unlocked from the factory providing ± 5% Error throughout the absorbance dynamic range.
- Accurate high concentration measurements — Absorbance measurements within 5% error up to 550 Abs (10 mm equivalent), up to 400 mg/mL IgG or 18,000 ng/μL.
- Quick, dilution free process — Take triplicate measurements on the pedestal in under 2 minutes.
- Enhanced calibration – bundled purchases get an additional calibration from the factory to calibrate the instrument for exceptional accuracy within the Acclaro Pro applications.
For research use only; not for diagnostic purposes.
Specifications
Specifications
| Product Line | NanoDrop |
| Type | ND UltraC FL and Acclaro Pro |
| For Use With (Equipment) | NanoDrop Ultra Spectrophotometers and Fluorometers |
| Includes | USB flash drive containing the Acclaro Pro software for use with the NanoDrop Ultra Spectrophotometers and Fluorometers NanoDrop UltraC FL |