Description
Certified standards for verifying instrument performance to factory specifications are available separately.
For use with NanoDrop Eight UV-Vis Spectrophotometer Solution with certified absorbance values. Use to create a record of correct photometric performance.
Contains: Two vials of CHEM-PV-1, two pastuer pipettes, and two 8-well PCR strip tubes.
Intended for single use only.
Specifications
Specifications
| Type | PV-8 Performance Verification |
| For Use With (Equipment) | NanoDrop Eight |