Learn More
Macherey-Nagel™ Nano Durasil-20 TLC Plate
Product Code. 10504922
Click to view available options
Dimensions (L x W):
10 x 10 cm
10 x 20 cm
Unit Size:
Pack of 25
Pack of 50
Product Code. 10504922
Brand: Macherey-Nagel™ 812011
Only null left
Specifications
Material Nano Durasil-20
Particle Size 2 to 10 μm
Pore Size 60 Å
Quantity 50 Pk.
Type TLC Plate
Pore Volume 0.75 mL/g
Specific Surface ∼500 m2/g
Thickness (Metric) 0.2 mm
Dimensions (L x W) 10 x 20 cm