Learn More
Macherey-Nagel™ Nano Durasil-20 TLC Plate
Click to view available options
Dimensions (L x W):
10 x 10 cm
10 x 20 cm
Unit Size:
Pack of 25
Pack of 50
Specifications
Specifications
| Material | Nano Durasil-20 |
| Particle Size | 2 to 10 μm |
| Pore Size | 60 Å |
| Quantity | 25 Pk. |
| Type | TLC Plate |
| Pore Volume | 0.75 mL/g |
| Specific Surface | ∼500 m2/g |
| Thickness (Metric) | 0.2 mm |
| Dimensions (L x W) | 10 x 10 cm |