missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Fmoc-L-glutamine, 95%, Thermo Scientific™

Product Code. 15435388
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15435388

Brand: Thermo Scientific Alfa Aesar H59159.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71989-20-3
Molecular Formula C20H20N2O5
Molecular Weight (g/mol) 368.39
MDL Number MFCD00037137
InChI Key IZKGGDFLLNVXNZ-UHFFFAOYNA-N
Synonym fmoc-gln-oh, nalpha-fmoc-l-glutamine, fmoc-l-gln-oh, fmoc-l-glutamine, n-fmoc-l-glutamine, fmoc-dap ac-oh, fmoc-gln, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-amino-5-oxopentanoic acid, 2s-4-carbamoyl-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, n alpha-fluorenylmethoxycarbonylglutamine
PubChem CID 2724775
SMILES NC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 224°C (decomposition)
Quantity 1 g
Beilstein 4722773
Optical Rotation −19° (c=1 in DMF)
IUPAC Name (2S)-5-amino-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxopentanoic acid
Formula Weight 368.38
Percent Purity 95%
Chemical Name or Material Nalpha-Fmoc-L-glutamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.