Learn More
Thermo Scientific™ Nalgene™ Torque Wrench Fittings for HDPE Closures
Product Code. 10190703
Click to view available options
Closure Size:
20 to 415 for serum vial
24 to 415 modified
28 to 415 modified
38 to 415 modified
38 to 430 modified
43 to 415 modified
53 to 415 modified
53B
83B
Unit Size:
Each
Product Code. 10190703
Brand: Thermo Scientific™ 21951028
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Choose these torque wrench fittings designed for specific HDPE closures to achieve accurate torque readings when applying closures onto bottles.
Choose Thermo Scientific™ Nalgene™ Torque Wrench Fittings for HDPE Closures to achieve accurate torque readings when applying closures onto bottles. Proper torquing of Nalgene closures is critical to leakproof† sealing.
  • Epoxy
  • Specifically designed for Nalgene HDPE closures
Compatible with:
Thermo Scientific Nalgene Torque Wrenches (2195-6169 and -6177) † The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Fittings
Closure Size 28 to 415 modified
Drive Size 1/4 in.
For Use With (Equipment) Nalgene Torque Wrenches (Cat. Nos. 2195-6169 and -6177)
Material Epoxy