Learn More
Thermo Scientific™ Nalgene™ Super Versi-Dry™ Surface Protectors
Product Code. 10661922
Change view
Click to view available options
Dimensions (L x W):
18 x 20 in.
18 x 40 in.
20 in. x 100 ft.
20 in. x 250 ft.
20 x 43 in.
40 in. x 200 ft.
Description:
250 ft. Dispenser Pk. Roll
Long Mat
Mat
Roll
Table Mat
Wide Roll
Unit Size:
4 rolls
61m
Each
Pack of 100
Pack of 150
Pack of 175
Pack of 350
Pack of 50
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Description Unit Size
10661922 20 in. x 250 ft. 250 ft. Dispenser Pk. Roll Each
10182361 18 x 20 in. Mat Pack of 350
10395951 18 x 40 in. Long Mat Pack of 175
10460733 20 x 43 in. Table Mat Pack of 150
10779441 20 in. x 100 ft. Roll 4 rolls
10789601 40 in. x 200 ft. Wide Roll 61m
10325861 18 x 20 in. Mat Pack of 100
10264461 18 x 40 in. Long Mat Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10661922 Supplier Thermo Scientific™ Supplier No. 7400000
  • ONLINE EXCLUSIVE PRICE
    266.00€
    239.99€ / Pack of 1
    Save 26.01€
    9% Off
In Stock
Add to basket
Edge
Add to basket
Absorb spills and cushion breakable items with long-lasting, waterproof, skid-resistant and chemical-resistant surface protectors.
Absorb spills and cushion breakable items with Thermo Scientific™ Nalgene™ Super Versi-Dry™ Surface Protectors. They are long-lasting, waterproof, skid-resistant and chemical-resistant. The cloth-like top layer will not fall apart even when wet.
  • Cushioned surface minimizes breakage of glass labware when dropped or tipped over
  • Sheets lie flat and are skid resistant
  • Low-pile flat surface provides stability for glassware that is likely to tip
  • Polyethylene backing resists harsh chemicals and protects benchtop from spills and corrosion
  • Super Versi-Dry quickly absorbs up to 1050mL/m2 to quickly absorb spills with thousands of Thermo Scientific Thirsty Cells
TRUSTED_SUSTAINABILITY
Specifications
Product Type Surface Protector
Absorbancy 1050 mL/m2
Length (English) 250 ft.
Length (Metric) 76 m
Width (English) 20 in.
Width (Metric) 51 cm
Dimensions (L x W) 20 in. x 250 ft.
Description 250 ft. Dispenser Pk. Roll
Quantity 1 Roll