Learn More
Thermo Scientific™ Nalgene™ Stopcocks
Product Code. 15223079
Click to view available options
Plug Bore:
2 mm
4 mm
Unit Size:
Each
Pack of 6
Product Code. 15223079
Brand: Thermo Scientific™ 64600004
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Develop in-house fluid transfer systems for accurate filling or dispensing. These stopcocks provide reliable, accurate liquid flow control.
Permit reliable, accurate liquid flow control in liquid handling systems by using Thermo Scientific™ Nalgene™ Stopcocks. They are ideal for use in developing in-house fluid transfer systems for one-way fluid path for accurate filling or dispensing. The two-way stopcocks feature serrated tubulation on each end.
  • Serrated tubulation on each end accepts 1/2″ through 5/16″ tubing
  • Polypropylene with Teflon™ TFE plug
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Type Stopcock
Plug Bore 4 mm
Material Polypropylene
No. per Pack 1
No. per Case 6
For Use With (Equipment) In-House Fluid Transfer System
Plug Material Polypropylene
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.