Learn More
Thermo Scientific™ Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box
Product Code. p-4520565 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
Cryogenic Box, 81-Ampule
For Use With (Equipment):
For Locator Jr. Plus
For: Locator 4 Plus
Locator Jr.
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds For Use With (Equipment) unitSize
10542144 Cryogenic Box, 81-Ampule Locator Jr. Each
12787065 Cryogenic Box, 81-Ampule For: Locator 4 Plus Each
10185433 Cryogenic Box, 81-Ampule For Locator Jr. Plus Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10542144 Supplier Thermo Scientific™ Supplier No. HR509X19A
Take advantage of efficient, high-volume storage with Thermo Scientific™ Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box.
Take advantage of efficient, high-volume storage with Thermo Scientific™ Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box.
  • Designed for long-term storage
  • Features an indexed frame and box organizational system
  • Unique hanging rack promotes easy sample retrieval
  • Suited for both manual and computerized inventory record-keeping methods
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
For Use With (Equipment) Locator Jr.
Holds Cryogenic Box, 81-Ampule
Type Locator™ Plus Cryogenic Rack