Learn More
Thermo Scientific™ Nalgene™ Heavy-Duty Rectangular LLDPE Tank with Covers
Product Code. p-4525591
Change view
Click to view available options
Capacity (Metric):
8 L
20 L
30 L
38 L
50 L
60 L
120 L
160 L
Width (Metric):
20 cm
25 cm
30 cm
45 cm
Length (Metric):
20 cm
25 cm
30 cm
45 cm
61 cm
Height (Metric):
20 cm
30 cm
36 cm
45 cm
61 cm
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Width (Metric) Length (Metric) Height (Metric) unitSize
10272701 8 L 20 cm 20 cm 20 cm Each
10324501 20 L 25 cm 25 cm 36 cm Each
10779041 30 L 30 cm 30 cm 30 cm Each
10109860 38 L 30 cm 45 cm 30 cm Each
10040671 50 L 30 cm 61 cm 30 cm Each
10080701 60 L 30 cm 45 cm 45 cm Each
10536551 120 L 45 cm 61 cm 45 cm Each
10204451 160 L 45 cm 61 cm 61 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10272701 Supplier Thermo Scientific™ Supplier No. 141000002
Only null left
Add to basket
Add to basket
Versatility at a lower cost than stainless-steel tanks.
Versatility at a lower cost than stainless-steel tanks. Impact- and abrasion-resistant Thermo Scientific™ Nalgene™ Heavy-Duty Cylindrical LLDPE Tanks withstand most acid, alcohols and bases. Offered in sizes from 19 to 208L.
Features:
  • Molded as one piece, no seams
  • Nominal wall thickness is 5/32 in. (3.97mm)
  • Stepped flange at top provides reinforcement and grip for lifting
  • Can be modified with factory-installed spigot
  • Not autoclavable
Note:
Tanks used at elevated temperatures or with high specific-gravity liquids should be externally supported.
TRUSTED_SUSTAINABILITY
Specifications
Length (English) 8 in.
Type Tank with Cover
Material LLDPE
Capacity (English) 2 gal.
Capacity (Metric) 8 L
Length (Metric) 20 cm
Width (English) 8 in.
Width (Metric) 20 cm
Height (English) 8 in.
Height (Metric) 20 cm
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Caution:
Plastic tanks are generally subject to more severe conditions that plastic labware; exposure is constant, stresses are greater and different classes and concentrations of chemicals are involved. Please pay special attention to chemical compatibility.