Learn More
Thermo Scientific™ Nalgene™ Cylindrical Polypropylene Tank with Cover
Product Code. p-4525599
Change view
Click to view available options
Capacity (Metric):
19 L
28 L
38 L
56.8 L
114 L
208 L
378 L
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10102211 19 L Each
10450913 28 L Each
10010601 38 L Each
10505791 56.8 L Each
10604602 114 L Each
10325801 208 L Each
10611722 378 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10102211 Supplier Thermo Scientific™ Supplier No. 112000005
Only null left
Add to basket
Add to basket
Resist stress cracking and increase tolerance to many organic chemicals with this polypropylene tank with Cover, an economic alternative to stainless-steel tanks.
Resist stress cracking and increase tolerance to many organic chemicals with the Thermo Scientific™ Nalgene™ Cylindrical Polypropylene Tank with Cover, an economic alternative to stainless-steel tanks.
Autoclavable cylindrical tanks are available in sizes ranging from 19L to 378L with a matching cover to significantly reduce evaporation and contamination.
Ordering Information:
Factory-installed spigots available. Nalgene Tank Liners, Cat. No. 343050, are sold separately.
Note:
Tanks used at elevated temperatures or with high specific-gravity liquids should be externally supported.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Outer 11 in.
Diameter (Metric) Outer 27.9 cm
Type Tank with Cover
Material Polypropylene
Capacity (English) 5 gal.
Capacity (Metric) 19 L
Height (English) 15 in.
Height (Metric) 38.1 cm
Can I buy a replacement lid for my Nalgene Cylindrical Polypropylene Tank with Cover?
Our Nalgene Cylindrical Polypropylene Tanks are molded as one piece and then cut to create the lid. As a result, we do not have replacement lids.
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. Plastic tanks are generally subject to more severe conditions than plastic labware; exposure is constant, stresses are greater and different classes and concentrations of chemicals are involved. Please pay special attention to chemical compatibility.