missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies NaïveCult™ Induction Kit for the generation of reset hPSCs
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
- Perform transgene-free chemical induction of reset human naïve embryonic stem (ES) and induced pluripotent stem (iPS) cells from the primed state using a defined cell culture medium
- Reset naïve human pluripotent stem cells (hPSCs) generated using this kit demonstrate uniformly domed, phase-bright colonies with refractive edges and maintain gene expression profiles associated with reset naïve hPSCs (Guo et al.) Stem Cell Reports, 2016; (Guo et al.) Development, 2017; Takashima et al.) Cell, 2014)
- Based on the formulation by (Guo et al.) (2017) and developed under license from the Cambridge Stem Cell Institute at the University of Cambridge, UK, this kit works robustly across multiple primed human ES and iPS cells maintained in mTeSR1 (Catalog #85850) or TeSR-E8 (Catalog #05990)
- Note that NaïveCult Induction Kit requires a histone deacetylase inhibitor (HDACi; either Sodium Butyrate [Catalog #72242] or VaLproic Acid [Catalog #72292]) that is sold separately
Specifications
Specifications
| Cell Type | Pluripotent Stem Cells |
| For Use With (Application) | Cell Culture, Expansion, Maintenance |
| Quantity | 1 Kit |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction