Learn More
Nabumetone, TRC
Product Code. 30481562
Change view
Click to view available options
Quantity:
50 mg
10 g
100 g
Unit Size:
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481562 10 g 10g
30466021 100 g -
30473677 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30481562 Supplier Toronto Research Chemicals Supplier No. TRCN20050010G
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 42924-53-8
Molecular Formula C15 H16 O2
Molecular Weight (g/mol) 228.29
Synonym Nabumetone, 4-(6-Methoxy-2-naphthalenyl)-2-butanone, 1-(2'-Methoxynaphth-6'-yl)butan-3-one, 4-(6-Methoxy-2-naphthyl)-2-butanone, Arthaxan, BRL 14777, Balmox, Consolan, Nabumeton, Nabumetone, Nabuser, Relafen, Relifen, Relifex
IUPAC Name 4-(6-methoxynaphthalen-2-yl)butan-2-one
SMILES COc1ccc2cc(CCC(=O)C)ccc2c1
Specifications
Quantity 10 g
InChI Formula InChI=1S/C15H16O2/c1-11(16)3-4-12-5-6-14-10-15(17-2)8-7-13(14)9-12/h5-10H,3-4H2,1-2H3
Formula Weight 228.115
Chemical Name or Material Nabumetone