Learn More
Nabertherm Muffle Furnaces
Nabertherm furnaces are perfect all-rounder for research and laboratory applications. They are manufactured from expertly finished, high quality materials and are easy to operate. The insulation out of non-classified fibre materials permits energysaving operation and fast heating times due to low heat storage and thermalconductivity.
Brand:  Nabertherm L-032K1RN1
Product Code. 15483392
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Heating from two sides by ceramic heating plates with integrated heating element which is safeguarded against fumes and splashing, and easy to replace
  • Insulation made of non-classified fibre material
  • Textured stainless steel sheet housing
  • Equipped with either a flap door (L) or lift door (LT)
  • Dual shell housing for low external temperatures and high stability
  • Adjustable air inlet integrated into the door, exhaust air outlet in the rear wall of furnace
  • Supplied with B410 controller with four programs and five freely programmable ramps, or C450 with 10 freely adjustable programs and 20 segments
  • Both controllers have integrated USB interface and offer plain text display in different languages
  • Alternative furnace sizes and temperature ranges are available on request