Learn More
Tocris Bioscience™ NAB 2
Protects against α-synuclein toxicity; promotes Rsp5/Nedd4-dependent endosomal transport
Brand:  Tocris Bioscience™ 5131/10
Encompass
Product Code. 15796496
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
NAB 2 protects against α-synuclein toxicity. NAB 2 reverses the α-synuclein-induced pathological phenotype in Parkinson's disease cortical neurons. NAB 2 promotes E3 ubiquitin ligase Rsp5/Nedd4-dependent endosomal transport.
Specifications
Specifications
>98%
10 mg