Learn More
Tocris Bioscience™ NAB 2
Protects against α-synuclein toxicity; promotes Rsp5/Nedd4-dependent endosomal transport
Brand: Tocris Bioscience™ 5131/10
This item is not returnable.
View return policy
Description
NAB 2 protects against α-synuclein toxicity. NAB 2 reverses the α-synuclein-induced pathological phenotype in Parkinson's disease cortical neurons. NAB 2 promotes E3 ubiquitin ligase Rsp5/Nedd4-dependent endosomal transport.Specifications
| >98% | |
| 10 mg |