Learn More
Macherey-Nagel™ N9 Polypropylene Red Screw Caps
Product Code. 14843532
Click to view available options
Color:
Red
Unit Size:
Pack of 100
Product Code. 14843532
Brand: Macherey-Nagel™ 702037
Only null left
Specifications
Color Red
Material Polypropylene, PTFE, Silicone
Septum Silicone white/PTFE red
Thickness (Metric) 1 mm
Closure Size N9