Learn More
Macherey-Nagel™ N9 Polypropylene Blue Screw Caps
Product Code. 10411708
Click to view available options
Septum:
Red Rubber/Clear FEP
White Silicone/Blue PTFE
White Silicone/Red PTFE
Unit Size:
Pack of 100
Product Code. 10411708
Brand: Macherey-Nagel™ 702732
Only null left
Add to basket
Add to basket
Seal vials with screw caps with a secure septum to receive needles. The Macherey-NagelTM N9 pollypropylene blue screw caps come with a choice of septa.
  • Screw closures N 9
  • Large range of screw closures with different septum materials and cap colors
  • Bonded screw caps (septa firmly connected with the cap; cannot be removed) – advantage: thick (blunt) HPLC needles cannot push the septum into the vial
  • Polymagnetic screw closure 702166 for CTC GC PAL
Specifications
Color Blue
Material FEP, Polypropylene, Rubber
Pre-slit No
Septum Red Rubber/Clear FEP
Thickness (Metric) 1 mm
Closure Size N9