Learn More
N6,N6,2'-O-Trimethyladenosine, TRC
Specifications
Specifications
| Chemical Name or Material | N6,N6,2'-O-Trimethyladenosine |
| CAS | 30891-53-3 |
| Molecular Formula | C13H19N5O4 |
| Quantity | 0.5 mg |
| Synonym | N,N-Dimethyl-2'-O-methyladenosine |
| InChI Formula | InChI=1S/C13H19N5O4/c1-17(2)11-8-12(15-5-14-11)18(6-16-8)13-10(21-3)9(20)7(4-19)22-13/h5-7,9-10,13,19-20H,4H2,1-3H3/t7-,9-,10-,13-/m1/s1 |
| SMILES | CN(C)C1=NC=NC2=C1N=CN2[C@H]3[C@@H]([C@@H]([C@H](O3)CO)O)OC |
| Molecular Weight (g/mol) | 309.32 |