missing translation for 'onlineSavingsMsg'
Learn More

N4BP3 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA556607

Additional Details : Weight : 0.02000kg

Product Code. 15794511

  • 514.00€ / 100µL
Estimated Shipment: 15-05-2024
to see stock.
Add to basket

Description

Description

Immunogen sequence: FSPEELRAAL AGSRGSRQPD GLLRKGLGQR EFLSYLHLPK KDSKSTKNTK RAPRNEPADY ATLYYREHSR AGDFSKTSLP ERGRFD Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 88%.

N4BP3, also named as KIAA0341, a Fezzin family member, during nerve cell development. It is crucial for axonal and dendritic branching in developing neurons. It binds the protein NEDD4 and interacts with 14-3-3 proteins. N4BP3 is present in early neurites, growth cones, axons and dendrites of developing primary hippocampal neurons.
Specifications

Specifications

N4BP3
Polyclonal
Unconjugated
N4BP3
C330016O10Rik; EMBL:ABA06436.1}; KIAA0341; LZTS4; N4bp3; N4BP3 {ECO:0000250; N4bp3 {ECO:0000312; Nedd4 binding protein 3; nedd4 binding protein 3 {ECO:0000312; NEDD4-binding protein 3; RGD:1308356}; RGD1308356; UniProtKB:O15049}
Rabbit
Antigen affinity chromatography
RUO
23138
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry, Western Blot
0.8 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O15049
N4BP3
Recombinant protein corresponding to Human N4BP3
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only