Learn More
N4-Hydroxycytosine (>90%), TRC
Product Code. 30404920
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30404920 50 mg 50mg
30403721 250 mg -
30406308 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30404920 Supplier Toronto Research Chemicals Supplier No. TRCH80507850MG
 Restricted Item
Estimated Shipment: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 20555-88-8
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.1
Synonym 6-(Hydroxyamino)-2(1H)-pyrimidinone
IUPAC Name 6-(hydroxyamino)-1H-pyrimidin-2-one
SMILES ONC1=CC=NC(=O)N1
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C4H5N3O2/c8-4-5-2-1-3(6-4)7-9/h1-2,9H,(H2,5,6,7,8)
Formula Weight 127.0382
Chemical Name or Material N4-Hydroxycytosine