missing translation for 'onlineSavingsMsg'
Learn More

N2-Acetyl-L-asparagine, TRC

Product Code. 30010456
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010456 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30010456 Supplier Toronto Research Chemicals Supplier No. TRCA168780500MG
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4033-40-3
Molecular Formula C6H10N2O4
Synonym N-Acetyl-L-asparagine, N-Acetylasparagine, N2-Acetylasparagine, NSC 186895alpha-N-Acetyl-L-asparagine;
InChI Formula InChI=1S/C6H10N2O4/c1-3(9)8-4(6(11)12)2-5(7)10/h4H,2H2,1H3,(H2,7,10)(H,8,9)(H,11,12)/t4-/m0/s1
SMILES CC(=O)N[C@@H](CC(=O)N)C(=O)O
IUPAC Name (2S)-2-acetamido-4-amino-4-oxobutanoic acid
Molecular Weight (g/mol) 174.15
Quantity 500 mg
Formula Weight 174.0641
Shipping Condition Room Temperature
Chemical Name or Material N2-Acetyl-L-asparagine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.