missing translation for 'onlineSavingsMsg'
Learn More

N1-tert-Butoxycarbonyl 1-(3-Methoxyphenyl)hydrazine, TRC

Product Code. 30345006
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30345006 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30345006 Supplier Toronto Research Chemicals Supplier No. TRCB690495100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N1-tert-Butoxycarbonyl 1-(3-Methoxyphenyl)hydrazine
CAS 380383-81-3
Molecular Formula C12H18N2O3
Quantity 100 mg
Synonym 1-(3-Methoxyphenyl)hydrazinecarboxylic Acid 1,1-Dimethylethyl Ester
InChI Formula InChI=1S/C12H18N2O3/c1-12(2,3)17-11(15)14(13)9-6-5-7-10(8-9)16-4/h5-8H,13H2,1-4H3
SMILES COc1cccc(c1)N(N)C(=O)OC(C)(C)C
IUPAC Name tert-butyl N-amino-N-(3-methoxyphenyl)carbamate
Molecular Weight (g/mol) 238.28
Formula Weight 238.1317
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.