Learn More
N1-(3-((1-Benzyl-1H-indazol-3-yl)oxy)propyl)-N1,N3,N3-trimethylpropane-1,3-diamine, TRC
Product Code. 30336047
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336047 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30336047 Supplier Toronto Research Chemicals Supplier No. TRCB1314952.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N1-(3-((1-Benzyl-1H-indazol-3-yl)oxy)propyl)-N1,N3,N3-trimethylpropane-1,3-diamine
CAS 1337966-15-0
Molecular Formula C23H32N4O
Quantity 2.5 mg
Synonym Benzydamine Hydrochloride Impurity D
InChI Formula InChI=1S/C23H32N4O/c1-25(2)15-9-16-26(3)17-10-18-28-23-21-13-7-8-14-22(21)27(24-23)19-20-11-5-4-6-12-20/h4-8,11-14H,9-10,15-19H2,1-3H3
SMILES CN(C)CCCN(C)CCCOc1nn(Cc2ccccc2)c3ccccc13
IUPAC Name N'-[3-(1-benzylindazol-3-yl)oxypropyl]-N,N,N'-trimethylpropane-1,3-diamine
Molecular Weight (g/mol) 380.53
Formula Weight 380.2576