missing translation for 'onlineSavingsMsg'
Learn More

N-Trifluoroacetamido-1,4-di-p-nitrobenzoyl Daunosamine, TRC

Product Code. 30457239
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457239 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457239 Supplier Toronto Research Chemicals Supplier No. TRCT78150010MG
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Trifluoroacetamido-1,4-di-p-nitrobenzoyl Daunosamine
CAS 52583-22-9
Molecular Formula C22H18F3N3O10
Quantity 10 mg
Synonym 2,3,6-Trideoxy-3-[(trifluoroacetyl)amino]-lyxo-hexopyranose 1,4-Bis(4-nitrobenzoate)
InChI Formula InChI=1S/C22H18F3N3O10/c1-11-18(38-20(30)13-4-8-15(9-5-13)28(34)35)16(26-21(31)22(23,24)25)10-17(36-11)37-19(29)12-2-6-14(7-3-12)27(32)33/h2-9,11,16-18H,10H2,1H3,(H,26,31)
SMILES CC1OC(CC(NC(=O)C(F)(F)F)C1OC(=O)c2ccc(cc2)[N+](=O)[O-])OC(=O)c3ccc(cc3)[N+](=O)[O-]
IUPAC Name [6-methyl-5-(4-nitrobenzoyl)oxy-4-[(2,2,2-trifluoroacetyl)amino]oxan-2-yl] 4-nitrobenzoate
Molecular Weight (g/mol) 541.39
Formula Weight 541.0944
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.