Learn More
N-tert-Butoxycarbonyl-3,4-dibenzyloxyphenylethylamine, TRC
Specifications
Specifications
| Chemical Name or Material | N-tert-Butoxycarbonyl-3,4-dibenzyloxyphenylethylamine |
| CAS | 300362-80-5 |
| Molecular Formula | C27H31NO4 |
| Quantity | 250 mg |
| Synonym | N-[2-[3,4-Bis(phenylmethoxy)phenyl]ethyl]-carbamic Acid 1,1-Dimethylethyl Ester |
| InChI Formula | InChI=1S/C27H31NO4/c1-27(2,3)32-26(29)28-17-16-21-14-15-24(30-19-22-10-6-4-7-11-22)25(18-21)31-20-23-12-8-5-9-13-23/h4-15,18H,16-17,19-20H2,1-3H3,(H,28,29) |
| SMILES | CC(C)(C)OC(=O)NCCc1ccc(OCc2ccccc2)c(OCc3ccccc3)c1 |
| IUPAC Name | tert-butyl N-[2-[3,4-bis(phenylmethoxy)phenyl]ethyl]carbamate |
| Molecular Weight (g/mol) | 433.54 |
| Formula Weight | 433.2253 |