missing translation for 'onlineSavingsMsg'
Learn More

N-[(S)-(+)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine, 97%, Thermo Scientific™

Product Code. 10081861 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10081861

Brand: Thermo Scientific Acros 332250050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 82717-96-2
Molecular Formula C15H21NO4
Molecular Weight (g/mol) 279.34
MDL Number MFCD00209976
InChI Key CEIWXEQZZZHLDM-AAEUAGOBSA-N
Synonym s-2-s-1-ethoxy-1-oxo-4-phenylbutan-2-yl amino propanoic acid, n-s-1-ethoxycarbonyl-3-phenylpropyl-l-alanine, unii-10sz4ij77b, n-1 s-ethoxycarbonyl-3-phenylpropyl-l-alanine, n-s-+-1-ethoxycarbonyl-3-phenylpropyl-l-alanine, 2s-2-2s-1-ethoxy-1-oxo-4-phenylbutan-2-yl amino propanoic acid, s,s-n-ethoxy carbonyl-3-phenylpropyl alanine, n-s-1-carboxyethyl-l-homophenylalanine ethyl ester, s,s-n-1-ethoxycarbonyl-3-phenylpropyl alanine
PubChem CID 5702571
SMILES CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@@H](C)C(O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 148°C to 153°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 5 g
IUPAC Name (2S)-2-{[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}propanoic acid
Formula Weight 279.34
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material N-[(S)-(+)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.