Learn More
n-Propyl-d7-benzene, CDN
Product Code. 30317821
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30317821
Brand: CDN CDND15690.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Propylbenzene-D7 (propyl-D7)
CAS 65087-58-3
Molecular Formula C9 D7 H5
Quantity 0.1 g
Synonym Benzene, propyl-d7- (9 CI), n-Propyl-d7-benzene, Propyl-d7-benzene, Propylbenzene-d7, Propylbenzene-D7 (propyl-D7)
InChI Formula InChI=1 S/C9H12/c1-2-6-9-7-4-3-5-8-9/h3-5,7-8 H,2,6H2,1H3/i1D3,2D2,6D2
SMILES [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])c1ccccc1
IUPAC Name 1,1,2,2,3,3,3-heptadeuteriopropylbenzene
Molecular Weight (g/mol) 127.235
Formula Weight 127.138 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity