missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Propyl 4-Hydroxybenzoate-2,3,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 4-Hydroxybenzoic-2,3,5,6-D4 Acid Propyl Ester |
| CAS | 1219802-67-1 |
| Molecular Formula | C10 H12 O3 |
| Quantity | 0.05 g |
| Synonym | 4-Hydroxybenzoic acid-propyl ester D4 |
| InChI Formula | InChI=1 S/C10H12O3/c1-2-7-13-10(12)8-3-5-9(11)6-4-8/h3-6,11 H,2,7H2,1H3/i3D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c(C(=O)OCCC)c([2 H])c([2 H])c1O |
| IUPAC Name | propyl 2,3,5,6-tetradeuterio-4-hydroxybenzoate |
| Molecular Weight (g/mol) | 184.2251 |
| Formula Weight | 184.1038 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction