Learn More
n-Propyl-1,1,2,2-d4 Alcohol, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1-Propanol-1,1,2,2-D4 |
| CAS | 70907-80-1 |
| Molecular Formula | C3 D4 H4 O |
| Quantity | 0.1 g |
| Synonym | 1-Propan-1,1,2,2-d4-ol (9 CI), 1,1,2,2-Tetradeuteropropanol, 1,1,2,2-Tetradeuteropropan-1-ol, Propyl-1,1,2,2-d4 alcohol, 1-Propanol-1,1,2,2-D4 |
| InChI Formula | InChI=1 S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3/i2D2,3D2 |
| SMILES | [2 H]C([2 H])(C)C([2 H])([2 H])O |
| IUPAC Name | 1,1,2,2-tetradeuteriopropan-1-ol |
| Molecular Weight (g/mol) | 64.12 |
| Formula Weight | 64.083 g/mol |
| Show More |